Learn More
Aviva Systems Biology Corp ANXA8L2 Antibody - N-terminal region (ARP36575_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36575_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANXA8L2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ANXA8L2. Alternate names: ANXA8, FLJ32754, bA145E20.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53054