Learn More
Medchemexpress LLC 2-Pyrimidinamine, 4-[4-(1H-imidazol-1-yl)phenyl]-N-2-propyn-1-yl- | 2234272-10-5 | 98.1% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY130530
Encompass_Preferred
AP-C5 is a selective inhibitor of cGMP-dependent protein kinase II (cGKII) with a pIC50 of 7.2. It can be used for research on diarrheal disease and also functions as a click chemistry reagent containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Selective inhibitor of cGMP-dependent protein kinase II (cGKII)
  • pIC50 of 7.2 for cGKII
  • Useful for diarrheal disease research
  • Click chemistry reagent with an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Potentiates cAMP signaling by PDE inhibition
  • Partially blocks heat-stable toxin (STa)-mediated short-circuit current (Isc) response in mouse ileum
Catalog No. 50-003-41731