Learn More
ABclonal Technology Phospho-AKT1-S473 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP0140500UL
Encompass_Preferred
This gene encodes a member of the AKT serine-threonine kinase family (AKT1), which is involved in cell signaling. AKT proteins contain a pleckstrin homology domain, kinase domain, and a regulatory domain. They are phosphorylated by PI3K and play a key role in regulating cell proliferation, survival, metabolism, and angiogenesis. Activated by phosphatidylinositol 3,4,5-trisphosphate (PIP3), AKT proteins regulate numerous cellular functions and are critical for tumor cell survival, inhibiting apoptosis and regulating the mTOR pathway. AKT1 mutations are associated with cancer types, including breast, colorectal, and ovarian cancers, as well as disorders like Proteus syndrome and Cowden syndrome. Multiple spliced variants of this gene exist.
Catalog No. 50-001-13290