Learn More
ABclonal Technology Phospho-Src-S17 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP05221000UL
Encompass_Preferred
This gene is highly similar to the v-src gene of Rous sarcoma virus. This proto-oncogene may play a role in the regulation of embryonic development and cell growth. The protein encoded by this gene is a tyrosine-protein kinase whose activity can be inhibited by phosphorylation by c-SRC kinase. Mutations in this gene could be involved in the malignant progression of colon cancer. Two transcript variants encoding the same protein have been found for this gene.
Catalog No. 50-001-10383