Learn More
ABclonal Technology Phospho-MAPKAPK-2/MK2-T334 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP05881000UL
Encompass_Preferred
This gene encodes a member of the Ser/Thr protein kinase family. This kinase is regulated through direct phosphorylation by p38 MAP kinase. In conjunction with p38 MAP kinase, this kinase is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation and cell proliferation. Heat shock protein HSP27 was shown to be one of the substrates of this kinase in vivo. Two transcript variants encoding two different isoforms have been found for this gene.
Catalog No. 50-001-13109