Learn More
ABclonal Technology Phospho-PAK2-S20 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP08031000UL
Encompass_Preferred
The p21 activated kinases (PAK) are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. The PAK proteins are a family of serine/threonine kinases that serve as targets for the small GTP binding proteins, CDC42 and RAC1, and have been implicated in a wide range of biological activities. The protein encoded by this gene is activated by proteolytic cleavage during caspase-mediated apoptosis, and may play a role in regulating the apoptotic events in the dying cell.
Catalog No. 50-001-10979