Learn More
ABclonal Technology Phospho-C/EBPB-T235 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP10551000UL
Encompass_Preferred
This intronless gene encodes a transcription factor that contains a basic leucine zipper (bZIP) domain. The encoded protein functions as a homodimer but can also form heterodimers with CCAAT/enhancer-binding proteins alpha, delta, and gamma. Activity of this protein is important in the regulation of genes involved in immune and inflammatory responses, among other processes. The use of alternative in-frame AUG start codons results in multiple protein isoforms, each with distinct biological functions.
Catalog No. 50-001-11849