Learn More
Alkali Scientific Alkali Scientific AP1096 IsoPure™ 96, Automated Purification System
SDP
Supplier:  Alkali Scientific RS1096
Encompass_Preferred
Automated System for Magnetic Bead-Based Purification Compatible with all popular magnetic bead kits Touch screen panel for on-board programming and operation Process up to 96 samples in deep-well plates 8-position carousel with 2 heating blocks Sample volume 50µl – 1000µl   The Alkali Scientific AP1096, IsoPure™ 96, Automated Purification System  is a high throughput, automated system that improves the efficiency and reproducibility of magnetic bead-based purification protocols. Capable of processing up to 96 samples at one time, the instrument is easy to set up and program. Mixing, magnetic bead transfer, washing, and elution steps are performed automatically. The purification process can be completed within 10 to 60 minutes depending on the type of reagent kit used and complexity of the program.
Catalog No. 50-302-9659
Add to Cart
Request a Quote