Learn More
ABclonal Technology Phospho-STING/TMEM173-S365 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP11991000UL
Encompass_Preferred
Enables 2,3-cyclic GMP-AMP binding activity, cyclic-di-GMP binding activity, and ubiquitin protein ligase binding activity. Involved in several processes, including defense response to other organism, macroautophagy, and positive regulation of interferon-beta production. Acts upstream of or within cellular response to interferon-beta, positive regulation of transcription by RNA polymerase II, and regulation of inflammatory response. Located in several cellular components, including autophagosome, perinuclear region of cytoplasm, and peroxisome. Is expressed in ductus deferens, epididymis, ileum, and prostate gland. Human ortholog(s) of this gene implicated in STING-associated vasculopathy with onset in infancy. Orthologous to human STING1 (stimulator of interferon response cGAMP interactor 1).
Catalog No. 50-001-12636