Learn More
ABclonal Technology Phospho-IP3R-S1756 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP12241000UL
Encompass_Preferred
This gene encodes an intracellular receptor for inositol 1,4,5-trisphosphate. Upon stimulation by inositol 1,4,5-trisphosphate, this receptor mediates calcium release from the endoplasmic reticulum. Mutations in this gene cause spinocerebellar ataxia type 15, a disease associated with an heterogeneous group of cerebellar disorders. Multiple transcript variants have been identified for this gene.
Catalog No. 50-001-11588