Learn More
ABclonal Technology Phospho-TDP-43/TARDBP-S409/S410 Rabbit pAb
SDP
Supplier:  ABclonal Technology AP1241500UL
Encompass_Preferred
HIV-1, the causative agent of acquired immunodeficiency syndrome (AIDS), contains an RNA genome that produces a chromosomally integrated DNA during the replicative cycle. Activation of HIV-1 gene expression by the transactivator Tat is dependent on an RNA regulatory element (TAR) located downstream of the transcription initiation site. The protein encoded by this gene is a transcriptional repressor that binds to chromosomally integrated TAR DNA and represses HIV-1 transcription. In addition, this protein regulates alternate splicing of the CFTR gene. A similar pseudogene is present on chromosome 20.
Catalog No. 50-001-12377