Learn More
Medchemexpress LLC AP1867-2-carboxymeth 25mg | 2230613-03-1 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11442025MG
Encompass_Preferred
AP1867-2-(carboxymethoxy) is a synthetic FKBP12F36V-directed ligand fragment used as the FKBP12-binding moiety in dTAG/PROTAC constructs for targeted protein degradation research.
  • Used to link FKBP12F36V to a CRBN ligand for dTAG-mediated degradation.
  • High purity suitable for research (HPLC purity reported as 99.88%).
  • Available as solid and as a DMSO solution for convenient formulation.
  • Offered in multiple pack sizes to support screening and scale-up.
  • Soluble in DMSO for solution preparation.
Catalog No. 50-002-29082