Learn More
Selleck Chemical LLC AP20187
SDP
Supplier:  Selleck Chemical LLC S848710MM/1ML
Encompass_Preferred
AP20187 (B/B Homodimerizer) is a chemical inducer of dimerization that activates FKBP-Casp8
Catalog No. 50-000-01837