Learn More
Medchemexpress LLC AP24600 | 1300690-48-5 | MFCD27980055 | 99.9% | 277.28 g/mol | C16H11N3O2 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW059917
AP24600 is an inactive metabolite of Ponatinib supplied as a solid for research use only. It is typically used as a reference standard and for in vitro biochemical and pharmacological testing.
- Purity 99.94%
- Molecular formula C16H11N3O2
- Molecular weight 277.28 g/mol
- Appearance off-white to light yellow solid
- Solubility 2 mg/mL in DMSO (7.21 mM)
- Storage store at room temperature; in solvent store at -80°C (2 years) or -20°C (1 year)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.