Learn More
Medchemexpress LLC HY-126124 5mg Medchemexpress, AP39 CAS:1429061-80-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1261245MG
Encompass_Preferred
Medchemexpress, HY-126124 5mg AP39 CAS:1429061-80-2 AP39 is a triphenylphosphonium derivatised anethole dithiolethione and mitochondria-targeting hydrogen sulfide (H2S) donor. AP39 increases intracellular H2S levels. AP39 exerts cytoprotective effects and maintains mitochondrial DNA integrity under oxidative stress conditions. AP39 protects against myocardial reperfusion injury and protects against Alzheimer's disease in mice model. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1092