Learn More
Hach Company APA6000 Alkalinity Reagent Set
Supplier:  Hach Company 6001000
Encompass
APA6000 Alkalinity Analyzer Reagent Set includes a 1-L bottle of each APA Alkalinity Reagent 1 (2826153) is an acid titrant that reacts with the alkalinity in the sample APA Alkalinity Reagent 2 (2696653) is an indicator used to determine the endpoints
Catalog No. 50-002-07020