Learn More
Medchemexpress LLC HY-16060 50mg , Apalutamide CAS:956104-40-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1606050MG
Encompass_Preferred
HY-16060 50mg Apalutamide CAS: 956104-40-8 Apalutamide (ARN-509) is a potent and competitive androgen receptor (AR) antagonist, binding AR with an IC50 of 16 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0965