Learn More
Medchemexpress LLC Apalutamide-d4 | 1638885-65-0 | 99.9% | 481.46 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY16060S5MG
Encompass_Preferred
Apalutamide-d4 is a deuterium labeled Apalutamide. Apalutamide is a potent and competitive androgen receptor (AR) antagonist, binding AR with an IC50 of 16 nM.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-94568