Learn More
Medchemexpress LLC Apararenone (MT-3995) | 945966-46-1 | 99.1% | 364.39 | 100 MG

Supplier: Medchemexpress LLC HY10900210MM/1ML
Apararenone is a novel non-steroidal mineralocorticoid receptor antagonist designed for the treatment of diabetic nephropathies and non-alcoholic steatohepatitis. This compound is provided as a white to off-white solid.
- Acts as a mineralocorticoid receptor antagonist
- Developed for diabetic nephropathies and non-alcoholic steatohepatitis
- Ships at room temperature
- Store powder at -20°C for up to 3 years or 4°C for up to 2 years
- Store in solvent at -80°C for up to 2 years or -20°C for up to 1 year
- Soluble in DMSO at concentrations ≥ 125 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.