Learn More
Apexbio Technology LLC Apatinib, 50mg. Cas: 1218779-75-9 MFCD: MFCD22420814. VEGFR2 inhibitor, orally bioavailable, selective
SDP
Supplier:  Apexbio Technology LLC B230350
Encompass_Preferred
Apatinib YN968D1is a potent inhibior of the VEGF signaling pathway with the IC50 value of 1nM for VEGFR-2 in in vitro enzyme experiments [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5341