Learn More
Aat Bioquest APC [Allophycocyanin]
SDP
Supplier:  Aat Bioquest 2555
Encompass_Preferred
Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga. Like other phycobiliproteins, APC is fluorescent, with an extremely high absorptivity and a high quantum efficiency.
Catalog No. 50-195-3184