Learn More
Medchemexpress LLC APC 366 10mg | 158921-85-8 | 10MG
SDP
Supplier:  Medchemexpress LLC HY10599910MG
Encompass_Preferred
APC 366 is a selective inhibitor of mast cell tryptase (Ki 7 1 M) APC 366 inhibits antigen-induced early asthmatic response (EAR) late asthmatic response (LAR) and bronchial hyperresponsiveness (BHR) in a sheep model of allergic asthma[1 [2
Catalog No. 50-002-44232