Learn More
Biolegend APC anti-human CD294 (CRTH2)
Supplier:  Biolegend 350110
Encompass_Preferred
APC anti-human CD294 (CRTH2) [BM16]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1390450