Learn More
Biolegend APC anti-human CD3
Supplier:  Biolegend 344811
Encompass_Preferred
APC anti-human CD3 [SK7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 tests
Catalog No. 50-168-306