Learn More
Biolegend APC anti-human CD4
Supplier:  Biolegend 317416
Encompass_Preferred
APC anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC9997972