Learn More
Biolegend APC anti-human CD49a
Supplier:  Biolegend 328314
Encompass_Preferred
APC anti-human CD49a [TS2/7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-207-5797