Learn More
Biolegend APC anti-human Notch 2
Supplier:  Biolegend 348305
Encompass_Preferred
APC anti-human Notch 2 [MHN2-25]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green; Apps: FC; Size: 25 tests
Catalog No. 50-168-467