Learn More
Biolegend APC anti-human CD186 (CXCR6)
Supplier:  Biolegend 356006
Encompass_Preferred
APC anti-human CD186 (CXCR6) [K041E5]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1231528