Learn More
Biolegend APC anti-human CD201 (EPCR)
Supplier:  Biolegend 351906
Encompass_Preferred
APC anti-human CD201 (EPCR) [RCR-401]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC0944901