Learn More
Biolegend APC anti-human CD223 (LAG-3)
Supplier:  Biolegend 369211
Encompass_Preferred
APC anti-human CD223 (LAG-3) [7H2C65]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1690915