Learn More
Biolegend APC anti-human CD25 [S20019C]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Supplier:  Biolegend 385605
Encompass_Preferred
APC anti-human CD25 [S20019C]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-253-0514