Learn More
Biolegend APC anti-human CD274 (B7-H1, PD-L1)
Supplier:  Biolegend 374514
Encompass_Preferred
APC anti-human CD274 (B7-H1, PD-L1) [MIH3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1888724