Learn More
ABclonal Technology APC anti-Human CD66a/CEACAM1
SDP
Supplier:  ABclonal Technology A26474100T
Encompass_Preferred
This gene encodes a member of the carcinoembryonic antigen (CEA) family part of the immunoglobulin superfamily Located in a 1 2 Mb cluster on chromosome 19 it includes CEA cell adhesion molecules pregnancy-specific glycoproteins and 11 pseudogenes Initially identified in liver bile ducts as biliary glycoprotein the protein is now known as a cell adhesion molecule found on leukocytes epithelia and endothelia It mediates both homophilic and heterophilic binding and is involved in tissue structure angiogenesis apoptosis tumor suppression metastasis and immune modulation Multiple transcript variants exist though not all are fully characterized
Catalog No. 50-000-12741