Learn More
Biolegend APC anti-human Lineage Cocktail (CD3/14/16/19/20/56)
Supplier:  Biolegend 348803
Encompass_Preferred
APC anti-human Lineage Cocktail (CD3/14/16/19/20/56) [UCHT1; HCD14; 3G8; HIB19; 2H7; HCD56]; Isotype: Mouse IgG1/Mouse IgG2b; Reactivity: Human; Apps: FC; Size: 50 tests
Catalog No. NC1724698