Learn More
Biolegend APC anti-mouse H-2Kd/H-2Dd
Supplier:  Biolegend 114714
Encompass_Preferred
APC anti-mouse H-2Kd/H-2Dd [34-1-2S]; Isotype: Mouse (C3H) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1698840