Learn More
Cytek Biosciences Inc APC aMu CD5 Anti Primary
Supplier: Cytek Biosciences Inc 200051U025
Product Name: APC Anti-Mouse CD5 (53-7.3)
Product Class: Antibody
Type: Primary
Product Name: APC Anti-Mouse CD5 (53-7.3)
Cytek Catalog Number: 20-0051-U025
Marker: CD5
Target Species: Mouse
Clonality: Monoclonal
Clone: 53-7.3
Host Species: Rat
Isotype: IgG2a, k
Conjugation: APC
Size: 25 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.