Learn More
Biolegend APC anti-mouse CD107a (LAMP-1)
Supplier:  Biolegend 121614
Encompass_Preferred
APC anti-mouse CD107a (LAMP-1) [1D4B]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-965