Learn More
Biolegend APC anti-mouse TCR Vβ5.1, 5.2
Supplier:  Biolegend 139506
Encompass_Preferred
APC anti-mouse TCR Vβ5.1, 5.2 [MR9-4]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-165-092