Learn More
Biolegend APC/Cyanine7 anti-human CD38
Supplier:  Biolegend 303534
Encompass_Preferred
APC/Cyanine7 anti-human CD38 [HIT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Horse, Cow (Bovine); Apps: FC; Size: 100 tests
Catalog No. 50-403-232