Learn More
Biolegend APC/Cyanine7 anti-human CD4
Supplier:  Biolegend 357416
Encompass_Preferred
APC/Cyanine7 anti-human CD4 [A161A1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1226542