Learn More
Biolegend APC/Cyanine7 anti-mouse CD11c
Supplier:  Biolegend 117324
Encompass_Preferred
APC/Cyanine7 anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC9908059