Learn More
Biolegend APC/FireTM 750 anti-mouse CD24
Supplier:  Biolegend 101839
Encompass_Preferred
APC/Fire™ 750 anti-mouse CD24 [M1/69]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-272-2813