Learn More
ABclonal Technology APC Rabbit anti- CD96/TACTILE
SDP
Supplier:  ABclonal Technology A26001200T
Encompass_Preferred
Acts upstream of or within negative regulation of interferon-gamma production negative regulation of natural killer cell cytokine production and response to lipopolysaccharide Predicted to be located in cytoplasm Is expressed in several structures including blood central nervous system ganglia heart and lung Human ortholog(s) of this gene implicated in C syndrome Orthologous to human CD96 (CD96 molecule)
Catalog No. 50-000-12696