Learn More
ABclonal Technology APC Rabbit anti-Human CD161
SDP
Supplier:  ABclonal Technology A26051100T
Encompass_Preferred
Natural killer (NK) cells are lymphocytes that mediate cytotoxicity and secrete cytokines after immune stimulation Several genes of the C-type lectin superfamily including the rodent NKRP1 family of glycoproteins are expressed by NK cells and may be involved in the regulation of NK cell function The KLRB1 protein contains an extracellular domain with several motifs characteristic of C-type lectins a transmembrane domain and a cytoplasmic domain The KLRB1 protein is classified as a type II membrane protein because it has an external C terminus
Catalog No. 50-000-10279