Learn More
ABclonal Technology APC Rabbit anti-Mouse CD103
SDP
Supplier:  ABclonal Technology A27134200T
Encompass_Preferred
Predicted to enable metal ion binding activity Predicted to act upstream of or within cell adhesion and integrin-mediated signaling pathway Located in external side of plasma membrane Is expressed in several structures including alimentary system genitourinary system lymph node nasal septum and nucleus pulposus Orthologous to human ITGAE (integrin subunit alpha E)
Catalog No. 50-000-11833