Learn More
ABclonal Technology APC Rabbit anti-Mouse CD11a
SDP
Supplier:  ABclonal Technology A27338100UG
Encompass_Preferred
Predicted to enable ICAM-3 receptor activity and cell adhesion molecule binding activity Involved in cell-cell adhesion Acts upstream of or within several processes including activated T cell proliferation positive regulation of T cell proliferation and positive regulation of calcium-mediated signaling Located in cell-cell junction external side of plasma membrane and immunological synapse Is expressed in heart hemolymphoid system and vault of skull Orthologous to human ITGAL (integrin subunit alpha L)
Catalog No. 50-000-09522