Learn More
ABclonal Technology APC Rabbit anti-Mouse IgG1 mAb
SDP
Supplier:  ABclonal Technology A25698200T
Encompass_Preferred
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response Mouse IgG1 is composed of two heavy chains and two light chains and it is involved in opsonization complement activation and antibody-dependent cell-mediated cytotoxicity It is also involved in the regulation of immune responses including the activation of T cells and the inhibition of B cell activation Mouse IgG1 is widely used in research and diagnostic applications including ELISA Western blotting and immunohistochemistry It is also used in therapeutic applications such as immunotherapy and vaccine development
Catalog No. 50-000-11848