Learn More
Medchemexpress LLC APCDD1 (adenomatosis polyposis coli down-regulated 1) human recombinant Fc-fusion protein | >95.0% | 5 UG
SDP
Supplier:  Medchemexpress LLC HYP761545UG
Encompass_Preferred
Recombinant human APCDD1 Fc-fusion protein produced in HEK293 cells and supplied lyophilized for research applications. The construct includes residues L27-G486 (partial) with a C-terminal human Fc tag and functions as a negative regulator of Wnt signaling, relevant to studies of hair loss and cancer biology.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • C-terminal human Fc tag facilitates detection and purification.
  • Lyophilized formulation from PBS for convenient storage and shipping.
  • Available in small microgram quantities suitable for assay development.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 85.7 kDa; sequence spans L27-G486 (partial).
  • Inhibits Wnt signaling and is implicated in hereditary hypotrichosis simplex.
Catalog No. 50-002-94237