Learn More
AB Sciex LLC APCI+ CALIBRTN SOLUTION 100ML
Supplier: AB Sciex LLC 4460131
The Calibration Solution For The Tripletof 5600 And Tripletof 4600 Systems. This Solution Is Intended For Use In Positive Ion Mode With The Duospray. Source Where The Calibration Solution Is Introduced Using The Apci Probe And The Sample Would Be Analyzed Using The Esi Probe.