Learn More
Selleck Chemical LLC Apcin 5mg
Supplier: Selleck Chemical LLC S96055MG
Apcin (APC inhibitor) is an inhibitor of the E3 ligase activity of the mitotic anaphase-promoting complex/cyclosome (APC/C) that binds to Cdc20 and competitively inhibits the ubiquitylation of D-box-containing substrates. *For Research & Development use only. Product is not intended for drug, household, or human consumption.